Learn More
Stat3 (pY705) Mouse, Alexa Fluor 647, Clone: 4/P-STAT3, BD
Mouse Monoclonal Antibody
Brand:  BD Biosciences 562071
 View more versions of this product
Product Code. 15841619
  • ONLINE EXCLUSIVE PRICE
    £970.00
    £957.82 / Each
    Save £12.18
    1% Off
Description
STAT (Signal transducer and activator of transcription) of Y705 in Stat3 occurs in response to growth factors and cytokines, and is essential for normal transcription activity.
The 4/P-STAT3 monoclonal antibody recognizes the phosphorylated Y705 of Stat3.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stat3 (pY705)
Monoclonal
Alexa Fluor 647
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Phosphorylated Human Stat3 Peptide
250 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
4/P-STAT3
Acute-phase response factor, APRF
Mouse
Affinity Purified
RUO
Human, Mouse
IgG2a κ