missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT2 [p Tyr689, p Tyr690] Antibody (1021D) [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB2890JF669
This item is not returnable.
View return policy
Description
STAT2 Monoclonal antibody specifically detects STAT2 in Human samples. It is validated for Flow Cytometry,Western Blot,Simple Western
Specifications
| STAT2 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| interferon alpha induced transcriptional activator, ISGF-3, MGC59816, p113, signal transducer and activator of transcription 2, signal transducer and activator of transcription 2, 113kD, signal transducer and activator of transcription 2, 113kDa, STAT113 | |
| Phosphopeptide containing the human STAT2 Y690 site | |
| 0.1 mL | |
| Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 6773 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, Western Blot, Simple Western | |
| 1021D | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction