Learn More
STAT1 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10144-2-AP-20UL
 View more versions of this product
Product Code. 16866333
  • £135.00 / 20µL
Description
This antibody is a rabbit polyclonal antibody raised against residues near the N terminus of human STAT1.
Signal transducer and transcription activator that mediates cellular responses to interferons (IFNs), cytokine KITLG/SCF and other cytokines and other growth factors. Following type I IFN (IFN-alpha and IFN-beta) binding to cell surface receptors, signaling via protein kinases leads to activation of Jak kinases (TYK2 and JAK1) and to tyrosine phosphorylation of STAT1 and STAT2. The phosphorylated STATs dimerize and associate with ISGF3G/IRF-9 to form a complex termed ISGF3 transcription factor, that enters the nucleus. ISGF3 binds to the IFN stimulated response element (ISRE) to activate the transcription of IFN-stimulated genes (ISG), which drive the cell in an antiviral state. In response to type II IFN (IFN-gamma), STAT1 is tyrosine- and serine-phosphorylated. It then forms a homodimer termed IFN-gamma-activated factor (GAF), migrates into the nucleus and binds to the IFN gamma activated sequence (GAS) to drive the expression of the target genes, inducing a cellular antiviral state. Becomes activated in response to KITLG/SCF and KIT signaling. May mediate cellular responses to activated FGFR1, FGFR2, FGFR3 and FGFR4.
Specifications
Specifications
STAT1
Polyclonal
Unconjugated
STAT1
ISGF 3, STAT1, STAT91
Rabbit
Antigen Affinity Chromatography
RUO
25124, 6772
-20°C
Liquid
Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry, Immunocytochemistry
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P42224
STAT1
STAT1 Fusion Protein Ag0199
20 μL
Primary
Human, Rat