missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT1, Rabbit anti-Human, Clone: 3E6, Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB19116.20uL
This item is not returnable.
View return policy
Specifications
| STAT1 | |
| Monoclonal | |
| Unconjugated | |
| In PBS (50% glycerol, 0.1% BSA, 0.02% sodium azide). | |
| DKFZp686B04100/ISGF-3/STAT91 | |
| Rabbit | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| 3E6 | |
| Flow Cytometry (1 ug/mL-0.001 ug/mL ) The optimal working dilution should be determined by the end user. | |
| signal transducer and activator of transcription 1, 91kDa | |
| STAT1 | |
| A synthetic phosphopeptide corresponding to residues surrounding Tyr701 of human phospho STAT1. | |
| RUO | |
| 6772 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction