Learn More
Stat1 (pY701) Mouse, Alexa Fluor 647, Clone: 4A, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 612597
Description
Stat (Signal transducer and activators of transcription) proteins are critical mediators of the biologic activity of cytokines, including interleukins, interferons, erythropoietin, and growth factors. Ligand-receptor interaction leads to activation of constitutively associated JAK family kinases and subsequent recruitment/activation of Stat proteins by tyrosine phosphorylation. Active Stat proteins then move to the nucleus to promote transcription of cytokine-inducible genes. Seven Stat proteins have been cloned, each of which is differentially expressed and/or activated in a cytokine-specific and cell type-specific manner. Stat1 and Stat2 are components of the ISGF3 (Interferon-Stimulated Gene Factor 3) complex, which is the primary transcription activator induced by the binding of the interferon to a specific cell-surface receptor. Stat1 has two alternatively spliced isoforms, 91kDa Stat1α and 84kDa Stat1β; Stat1α has 38 additional C-terminal amino acids. In response to the binding of IFNα, IFNγ, EGF, PDGF, or CSF-1 to their respective receptors, the Stat1 subunits become tyrosine-phosphorylated at Y701, and the complex is translocated to the nucleus. This results in the formation of an active complex that includes the DNA-binding p48 subunit. This complex is responsible for modulating the transcription of the interferon-stimulated genes (ISGs). The 4a monoclonal antibody recognizes the phosphorylated Y701 in Stat1α and Stat1β.
Flow Cytometry (Intracellular Staining)
Specifications
| Stat1 (pY701) | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Phosphorylated Human Stat1 Peptide | |
| 50 Tests | |
| Cell Biology | |
| Human, Mouse | |
| IgG2a |
| Flow Cytometry | |
| 4A | |
| Signal Transducer and Activator of Transcription-1; STAT91; ISGF-3; CANDF7 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.