missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ STAT1 Protein

Product Code. p-5355370 Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
0.25 mg
Unit Size:
0.05mg
0.25mg
This item is not returnable. View return policy

Product Code. 18262675

Brand: Novus Biologicals™ NBP1483310.05MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-712 of Human STAT1 The Recombinant Human STAT1 Protein is derived from E. coli. The Recombinant Human STAT1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_009330
Concentration 0.5mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 0.1M NaCl, 1mM DTT, 10% glycerol
Gene ID (Entrez) 6772
Molecular Weight (g/mol) 85.2kDa
Purification Method Protein
Quantity 0.05 mg
Source Recombinant human STAT1 protein, fused to His-tag at N-terminus, expressed in E. coli
Immunogen STAT1, 1-712aa, Human, His tag, E. coli (NP_009330).
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name STAT1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.