missing translation for 'onlineSavingsMsg'
Learn More

STAT1 Antibody (OTI15H3), DyLight 594, Novus Biologicals™

Product Code. 18420087 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420087 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420087 Supplier Novus Biologicals Supplier No. NBP274377DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

STAT1 Monoclonal antibody specifically detects STAT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAT1
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI15H3
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias DKFZp686B04100, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human STAT1 expression vector.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Myeloid derived Suppressor Cell, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6772
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.