Learn More
Invitrogen™ STAT Family Activation Human InstantOne™ ELISA Kit
Description
Includes
- User Manual
- InstantOne™ Assay Plate - 96 well (sealed)
- Cell Lysis Buffer (5X)
- Enhancer Solution
- Wash Buffer (10X)
- phospho-STAT1 (Tyr701) Capture Antibody Reagent
- phospho-STAT3 (Tyr705) Capture Antibody Reagent
- phospho-STAT5A/B (Tyr694/699) Capture Antibody Reagent
- phospho-STAT1 (Tyr701) Detection Antibody Reagent
- phospho-STAT3 (Tyr705) Detection Antibody Reagent
- phospho-STAT5A/B (Tyr694/699) Detection Antibody Reagent
- Positive Control Cell Lysate (lyophilized)
- Detection Reagent (TMB Substrate)
- Stop Solution
- Plate Seal
The InstantOne™ ELISA is specifically engineered for accurate measurement of phosphorylated human STAT1 (Tyr701), STAT3 (Tyr705), and STAT5A/B (Tyr694/699) in cell lysates. Detection of mouse and rat phosphorylated STAT5A/B is also expected. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.
Specifications
Specifications
| Accession Number | P40763,P42224,P42229,P51692 |
| Conjugate | Biotin |
| Product Type | ELISA Kit |
| Sample Type | Cell Lysates |
| Target Species | Human |
| For Use With (Application) | ELISA |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 6772, 6774, 6776, 6777 |
| Gene Symbol | ADMIO,ADMIO1,APRF,HIES,CANDF7,IMD31A,IMD31B,IMD31C,ISGF-3,STAT91,MGF,STAT5 |
| Kit Contents | Capture Antibody, Detection Antibody Reagent, InstantOne™ Assay Plate, Cell Lysis Buffer, Enhancer Solution, Wash Buffer (10X), Substrate Reagent (TMB), Stop Solution, Positive Control Cell Lysate (lyophilized), Plate seal |
| Show More |
For Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.