Learn More
Manutan Starter workbench with 30mm vinyl top, 2000mm x 1400mm
Product Code. 12861498
Encompass_Preferred
Product Code. 12861498
Brand: Manutan A349277
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Add workbenches in 2m widths and accessories as required. Heavy duty modular design, the framework is constructed from fabricated steel section with welded leg supports. Bolt-in multi-position cross rails which can be assembled in a variety of combinations to suit individual applications. Base shelves optional, made from 25mm plain cut MDF. Base shelf load capacity 250kg, upper shelf load capacity 75kg. Supplied flat-packed, larger units have sectional worktops for ease of handling and are reversible to eliminate worn areas.
Dimensions (H x W x D): 840mm x 2000mm x 1400mm