missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Starter Kit for MethoCult™ H4534 Classic Without EPO

Product Code. 15509535
Encompass
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15509535

Brand: STEMCELL Technologies 04564

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Complete kit for hematopoietic CFU assays

  • The Starter Kit for MethoCult™ H4534 Classic without EPO (MethoCult™ GF H4534) is recommended for laboratories in the initial stages of establishing procedures for assessing human hematopoietic progenitor cells using colony-forming unit (CFU) assays
  • These products support the growth of clonogenic hematopoietic progenitor cells from human bone marrow, peripheral blood, cord blood, leukapheresis products, and purified progenitor-enriched cells
  • H4534 without EPO is a complete methylcellulose-based medium that supports the growth of granulocyte-macrophage progenitors (CFU-GM, CFU-G and CFU-M)
  • Each kit contains instructional materials in addition to all reagents and materials necessary to perform 24 duplicate assays

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells
For Use With (Application) Cell Culture, Colony Assay, Functional Assay
Quantity 1 Kit
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.