missing translation for 'onlineSavingsMsg'
Learn More
Learn More
START domain containing 7 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97964DL594
This item is not returnable.
View return policy
Description
START domain containing 7 Polyclonal specifically detects START domain containing 7 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| START domain containing 7 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Gestational trophoblastic tumor protein 1, GTT1stAR-related lipid transfer protein 7, mitochondrial, StARD7, StAR-related lipid transfer (START) domain containing 7, START domain containing 7, START domain-containing protein 7 | |
| Produced in rabbits immunized with E. coli-derived Human START domain containing 7 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 56910 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction