missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Stargazin Polyclonal antibody specifically detects Stargazin in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Stargazin |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 5 - 10 ug/ml |
| Formulation | PBS |
| Gene Alias | calcium channel, voltage-dependent, gamma subunit 2, FLJ41437, MGC138502, MGC138504, Neuronal voltage-gated calcium channel gamma-2 subunit, stargazin, voltage-dependent calcium channel gamma-2 subunit |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide derived from the rat calcium channel gamma2 subunit conjugated to KLH (Uniprot: Q9Y698) |
| Purification Method | Ammonium sulfate precipitation |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?