missing translation for 'onlineSavingsMsg'
Learn More

IBA Solutions For Life Science StarGate™ Upstream Fusion Vector pNFUSE-IBA

Product Code. 15693718 Shop All IBA Lifesciences Products
Click to view available options
Vector:
pNFUSE-IBA w/ IRES11
pNFUSE-IBA w/ LINK11
pNFUSE-IBA w/ LINK12
pNFUSE-IBA w/ SD11
Unit Size:
5µg
This item is not returnable. View return policy

Product Code. 15693718

Brand: IBA Solutions For Life Science 51631001

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For upstream positioning of an inserted gene in the final Fusion Donor vector when linking two or more genes cloned in separate Donor Vectors using the StarGate™ transfer cloning system.

StarGate™ offers a simple cloning procedure for rapid and highly efficient subcloning of an arbitrary gene of interest (GOI) for use in functional expression, purification, detection, and immobilization or separation of recombinant proteins.

  • Minimal extra modification of the gene due to short combinatorial sites
  • Inherent high-level cloning efficiency due to a directed reaction (no equilibrium)
  • Genes are cloned in a Donor Vector and then parallel-transfered into Acceptor Vectors that provide different genetic surroundings
  • Easily connect two or more genes that are cloned in separate Donor Vectors using Fusion Vectors that are assembled by use of a one-tube reaction
    • The GOI to be positioned upstream is transferred into a pNFUSE-IBA derivative, specifying the intergenic region (IR):
      • IRES11: Provides internal ribosomal entry site (IRES) from encephalomyocarditis virus for separate cap-independent translation of downstream cistron in mammalian cells
      • SD11: IR includes a Shine Dalgarno (SD) sequence for translation of downstream cistron at comparable level to the upstream cistron in bacterial cells
      • LINK11: Genes of interest are fused by a sequence encoding the 10 amino acid linker GlySerGlyGlyGlySerGlyGlyGlySer
      • LINK12: Genes of interest are fused by a sequence encoding the 15 amino acid linker GlySerGlyGlySerGlyGlyGlyGlySerGlyGlyGlyGlySer
    • The GOIs to be positioned downstream are transferred into a pCFUSE-IBA11 vector (identical, irrespective of IR)

Specifications

Format Solution, liquid
For Use With (Equipment) Stargate™ system derivatives
Description Resistance: ampicillin
Includes IR: Internal ribosomal entry site (IRES) from encephalomyocarditis virus
pH 8
Product Type Fusion Vector
Storage Buffer 20μL TE buffer: 10mM Tris-HCl, 1mM EDTA per 5μL pCFUSE-IBA11
Content And Storage 2°C to 8°C for frequent usage, -20°C for long-term storage
Concentration 250 ng/μL
Quantity 5 μg
Vector pNFUSE-IBA w/ IRES11
Color Clear
For Use With (Application) Upstream positioning, fusing cloned genes of interest
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.