missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences StarGate pYSG-IBA Vectors for Yeast Expression

Designed for high-level expression of recombinant proteins in Yeast by introducing the gene of interest under the control of the Cu++-inducible CUP1 promoter.

Brand:  IBA Lifesciences 5-4643-001

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 15673808

  • £229.00 / 5µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

pYSG-IBA yeast expression vectors

The pYSG-IBA vector allows the high-level expression of proteins in yeast.

  • The vector carries the copper inducible promoter (CUP1) for controlled high-level expression.
  • The expressed recombinant protein will be localized in the cytoplasm.

 

  • High-level expression in yeast with high copy number
  • Favors correct protein folding and the production of soluble proteins - inclusion bodies rarely form
  • Tightly regulated expression due to Cu++-inducible CUP1 promoter
  • Ampicillin resistance gene and markers Leu2-d and URA3 in every vector
  • Leu2-d auxotrophy marker for selection to increase plasmid copy number during expression
  • URA3 auxotrophy marker for selection after transformation

Please note that cloning into pYSG-IBA Acceptor Vectors compulsorily requires the restriction enzyme Esp3I since no other MCS for the integration of a gene of interest is available. In addition to the direct cloning of the gene of interest into pYSG-IBA vectors with Esp3I, another option via a so-called Entry-Vector is possible.

Specifications

Specifications

Liquid
pYSG-IBA 43
leu2-d and URA3
CUP1
20 μL TE buffer: 10 mm Tris-HCl, 1 mm EDTA / 5 μg
His Tag (6x) for N-terminal and Strep-tag for C-terminal
5 μg
Cloning Genes of Interest for Use in Functional Expression, Purification, Detection, Immobilization or Separation of Recombinant Proteins
StarGate™ System, Yeast Host Cells
Copper sulfate
8
Acceptor Vector for yeast expression
2°C to 8°C for frequent usage, -20°C for long-term storage
250 ng/μL
Clear
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers