missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences StarGate pYSG-IBA Vectors for Yeast Expression

Product Code. 15653808 Shop All IBA Lifesciences Products
Click to view available options
Vector:
pYSG-IBA 103
pYSG-IBA 105
pYSG-IBA 123
pYSG-IBA 143
pYSG-IBA 145
pYSG-IBA 162
pYSG-IBA 164
pYSG-IBA 167
pYSG-IBA 168
pYSG-IBA 23
pYSG-IBA 25
pYSG-IBA 3
pYSG-IBA 33
pYSG-IBA 43
pYSG-IBA 45
pYSG-IBA 5
pYSG-IBA 62
pYSG-IBA 63
pYSG-IBA 64
pYSG-IBA 65
pYSG-IBA wt1
Unit Size:
5µg
This item is not returnable. View return policy

Product Code. 15653808

Brand: IBA Lifesciences 54625001

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed for high-level expression of recombinant proteins in Yeast by introducing the gene of interest under the control of the Cu++-inducible CUP1 promoter.

pYSG-IBA yeast expression vectors

The pYSG-IBA vector allows the high-level expression of proteins in yeast.

  • The vector carries the copper inducible promoter (CUP1) for controlled high-level expression.
  • The expressed recombinant protein will be localized in the cytoplasm.

 

  • High-level expression in yeast with high copy number
  • Favors correct protein folding and the production of soluble proteins - inclusion bodies rarely form
  • Tightly regulated expression due to Cu++-inducible CUP1 promoter
  • Ampicillin resistance gene and markers Leu2-d and URA3 in every vector
  • Leu2-d auxotrophy marker for selection to increase plasmid copy number during expression
  • URA3 auxotrophy marker for selection after transformation

Please note that cloning into pYSG-IBA Acceptor Vectors compulsorily requires the restriction enzyme Esp3I since no other MCS for the integration of a gene of interest is available. In addition to the direct cloning of the gene of interest into pYSG-IBA vectors with Esp3I, another option via a so-called Entry-Vector is possible.

Specifications

Format Liquid
For Use With (Equipment) StarGate™ System, Yeast Host Cells
Inducing Agent Copper sulfate
Marker Gene leu2-d and URA3
pH 8
Promoter CUP1
Product Type Acceptor Vector for yeast expression
Storage Buffer 20 μL TE buffer: 10 mm Tris-HCl, 1 mm EDTA / 5 μg
Content And Storage 2°C to 8°C for frequent usage, -20°C for long-term storage
Protein Tag GST Tag for N-terminal
Concentration 250 ng/μL
Quantity 5 μg
Vector pYSG-IBA 25
Color Clear
For Use With (Application) Cloning Genes of Interest for Use in Functional Expression, Purification, Detection, Immobilization or Separation of Recombinant Proteins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.