missing translation for 'onlineSavingsMsg'
Learn More

STARD7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen STARD7
Concentration 0.35 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16854974
View Documents
Proteintech
15689-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16844974
View Documents
Proteintech
15689-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Although the function of this gene is not known, its existence is supported by mRNA and EST data. The predicted gene product contains a region similar to the STAR-related lipid transfer (START) domain, which is often present in proteins involved in the cell signaling mediated by lipid binding. Alternatively spliced transcript variants have been described, although some transcripts occur only in cancer cell lines.
Specifications

Specifications

STARD7
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Rat, Mouse, Human
Q8R1R3, Q9NQZ5
296128, 56910, 99138
STARD7 Fusion Protein Ag8224
Primary
-20°C
STARD7
0.35 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
GTT1, STARD7
STARD7
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.