Learn More
STARD5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559241
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: REGNGVSVSW RPSVEFPGNL YRGEGIVYGT LEEVWDCVKP AVGGLRVKWD ENVTGFEIIQ SITDTLCVSR TSTPS Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 84%.
May be involved in the intracellular transport of sterols or other lipids. May bind cholesterol or other sterols.Specifications
STARD5 | |
Polyclonal | |
Unconjugated | |
STARD5 | |
18B7-T7(GS); 2310058G22Rik; AI607880; D7Ertd152e; RGD1561783; StAR related lipid transfer domain containing 5; Stard5; StAR-related lipid transfer (START) domain containing 5; StAR-related lipid transfer domain containing 5; stAR-related lipid transfer protein 5; StAR-related protein 1-4E; StAR-related protein p3-15E/p3-16E; START domain containing 5; START domain-containing protein 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80765 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NSY2 | |
STARD5 | |
Recombinant protein corresponding to Human STARD5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only