missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STARD5 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94749-0.1ml
This item is not returnable.
View return policy
Description
STARD5 Polyclonal antibody specifically detects STARD5 in Human samples. It is validated for Western Blot
Specifications
| STARD5 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| MGC10327, StARD5, StAR-related lipid transfer (START) domain containing 5, stAR-related lipid transfer protein 5, START domain containing 5, START domain-containing protein 5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-213 of human STARD5 (NP_871629.1). MDPALAAQMSEAVAEKMLQYRRDTAGWKICREGNGVSVSWRPSVEFPGNLYRGEGIVYGTLEEVWDCVKPAVGGLRVKWDENVTGFEIIQSITDTLCVSRTSTPSAAMKLISPRDFVDLVLVKRYEDGTISSNATHVEHPLCPPKPGFVRGFNHPCGCFCEPLPGEPTKTNLVTFFHTDLSGYLPQNVVDSFFPRSMTRFYANLQKAVKQFHE | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 80765 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction