missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STARD4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£199.00 - £255.00
Specifications
| Antigen | STARD4 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:500-1:2000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18666915
|
Novus Biologicals
NBP2-99102-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18625135
|
Novus Biologicals
NBP2-99102-50ul |
50 μg |
£199.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
STARD4 Polyclonal antibody specifically detects STARD4 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| STARD4 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| StARD4, StAR-related lipid transfer (START) domain containing 4, stAR-related lipid transfer protein 4, START domain containing 4 sterol-regulated, START domain-containing protein 4, sterol regulated | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human STARD4. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 μm filtered solution in PBS | |
| 134429 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title