missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STARD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-599
This item is not returnable.
View return policy
Description
STARD3 Polyclonal specifically detects STARD3 in Rat samples. It is validated for Western Blot.
Specifications
| STARD3 | |
| Polyclonal | |
| Unconjugated | |
| PBS and 1 mg/ml BSA with 0.05% Sodium Azide | |
| STARD3 | |
| Recombinant MLN64 protein. | |
| 100 μg | |
| Breast Cancer, Cancer, Lipid and Metabolism | |
| 10948 | |
| Rat | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1 ug/ml | |
| CAB1, es64, Metastatic lymph node gene 64 protein, metastatic lymph node protein 64, MLN 64, MLN64FLJ41370, Protein CAB1, StARD3, StAR-related lipid transfer (START) domain containing 3, stAR-related lipid transfer protein 3, START domain containing 3, START domain-containing protein 3, steroidogenic acute regulatory protein related | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| MLN 64 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction