missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STARD13 Polyclonal Antibody

STARD13 Polyclonal Antibody
Brand: Thermo Scientific PA558831
This item is not returnable.
View return policy
Description
STARD13 Polyclonal Antibody for IHC
Specifications
| STARD13 | |
| Polyclonal | |
| Unconjugated | |
| STARD13 | |
| 46H23.2; ARHGAP37; deleted in liver cancer 2 protein; DLC2; DLC-2; GT650; LINC00464; long intergenic non-protein coding RNA 464; Rho GTPase activating protein on chromosome 13q12; Rho GTPase-activating protein; serologically defined colon cancer antigen 13; serologically defined colon cancer antigen 28; STA13; StAR related lipid transfer domain containing 13; STARD13; StAR-related lipid transfer (START) domain containing 13; StAR-related lipid transfer domain containing 13; stAR-related lipid transfer protein 13; START domain-containing protein 13 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90627 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y3M8 | |
| STARD13 | |
| Recombinant protein corresponding to Human STARD13. Recombinant protein control fragment (Product #RP-97949). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction