missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STARD10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17048-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody detected duplicate bands between 35-40 kDa in MCF7 and HepG2 cells. The similar expression pattern of STARD10 was also observed by other scientists, while the exact reason is not clear.
STARD10 gene ontology annotations related to this gene include bile acid secretion; positive regulation of peroxisome proliferator activated receptor signaling pathway.Specifications
| STARD10 | |
| Polyclonal | |
| Unconjugated | |
| STARD10 | |
| Antigen NY CO 28, CGI 52, NY CO 28, PCTP L, PCTP like protein, PCTP2, SDCCAG28, STARD10 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10809, 293150, 56018 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9JMD3, Q9Y365 | |
| Stard10 | |
| STARD10 Fusion Protein Ag10705 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction