missing translation for 'onlineSavingsMsg'
Learn More

STARD10 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£157.00 - £383.00

Specifications

Antigen STARD10
Concentration 1.0 mg/mL
Dilution Western Blot 1 - 2 μg/ml, Immunohistochemistry 1:200 - 1:400, Immunocytochemistry/ Immunofluorescence 1 - 3 μg/ml, Immunohistochemistry-Paraffin 1:200 - 1:400
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18666381
View Documents
Novus Biologicals
NBP2-78818
0.1 mg
£383.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18684829
View Documents
Novus Biologicals
NBP2-78818SS
0.025 mg
£157.00
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

STARD10 Polyclonal antibody specifically detects STARD10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

STARD10
Western Blot 1 - 2 μg/ml, Immunohistochemistry 1:200 - 1:400, Immunocytochemistry/ Immunofluorescence 1 - 3 μg/ml, Immunohistochemistry-Paraffin 1:200 - 1:400
Polyclonal
Purified
RUO
PBS
10809
IgG
Immunogen affinity purified
1.0 mg/mL
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse, Rat
Antigen NY-CO-28, CGI-52, MGC14401, NY-CO-28, PCTP2, PCTP-L, PCTP-like protein, Serologically defined colon cancer antigen 28SDCCAG28, StARD10, StAR-related lipid transfer (START) domain containing 10, StAR-related lipid transfer protein 10, START domain containing 10, START domain-containing protein 10
Full length recombinant human STARD10 protein (amino acids 1-291) [UniProt Q9Y365]
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.