missing translation for 'onlineSavingsMsg'
Learn More

Starch, soluble, ACS (for iodometry)

Product Code. 11384227
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384227 1 kg 1kg
11394227 50 g 50g
11374227 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11384227 Supplier Thermo Scientific Alfa Aesar Supplier No. 036703.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For iodometry | CAS: 9005-84-9 | C12H22O11 | 342.297 g/mol

In food industry, starch is used as a food preservative. Starch has extensive industrial use as an adhesive or bonding agent. It is also used in paper industry. Starch is used in the procedure to manufacture gypsum wallboards. Starch used in the laundry, also known as clothing starch. Glucose derived from starch may be fermented further to make ethanol or biofuel. Starch is also employed in the petroleum exploration industry. It is interesting to note that starch may be used to make hydrogen by making usage of enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-84-9
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.297
MDL Number MFCD00082026
InChI Key GUBGYTABKSRVRQ-ASMJPISFSA-N
Synonym alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
PubChem CID 439341
ChEBI CHEBI:18167
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O

Specifications

Physical Form Powder
Melting Point 256°C to 258°C (decomposition)
Merck Index 14,8799
Solubility Information Soluble in hot water.
Quantity 1 kg
Grade ACS
Odor Odorless
Chemical Name or Material Starch, Soluble

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.