missing translation for 'onlineSavingsMsg'
Learn More

Starch, MP Biomedicals

Product Code. 11495692 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 lb.
100 lb.
Unit Size:
100lb
25lb
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495692 25 lb. 25lb
11405702 100 lb. 100lb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495692 Supplier MP Biomedicals Supplier No. 0290295625
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Approx. 75% amylopectin, 25% amylose

Chemical Identifiers

CAS 9005-84-9
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.297
InChI Key GUBGYTABKSRVRQ-ASMJPISFSA-N
Synonym alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
PubChem CID 439341
ChEBI CHEBI:18167
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O

Specifications

Quantity 25 lb.
Physical Form Fine Powder
Chemical Name or Material Starch, From Corn
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.