missing translation for 'onlineSavingsMsg'
Learn More

Starch indicator solution 1%, w/v aqueous solution (for iodometric titrations), Thermo Scientific™

For iodometric titrations

Brand:  Thermo Scientific Alfa Aesar 047209.K7

 View more versions of this product

Product Code. 15457475

  • £58.20 / 4 L
 Restricted Item
Estimated Shipment: 27-08-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Starch indicator solution 1%
9005-84-9
MFCD00082026
alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
GUBGYTABKSRVRQ-ASMJPISFSA-N
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
439341
w/v aqueous solution for iodometric titrations
C12H22O11
14,8799
4L
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O
342.297
CHEBI:18167
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

RUO – Research Use Only