Learn More
Starch indicator solution 1%, Acculute Standard Volumetric Solution, Final Volume 1L, Thermo Scientific Chemicals
For Iodometric Titrations
Brand: Thermo Scientific Alfa Aesar 088632.LQ
71.25 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Code : NEW
Additional Details : CAS Number : 9005-25-8 Weight : 1.00000kg
Description
Starch indicator 1% solution is used as an indicator for redox titrations. It is also used in the detection of end point in iodometric titration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Starch indicator solution | |
9005-84-9,7732-18-5 | |
MFCD00082026 | |
1 U | |
17mm Hg at 20°C |
Liquid,For iodometric titrations | |
100% | |
Miscible with water. | |
Starch: 20%; Water: 80% |
RUO – Research Use Only