missing translation for 'onlineSavingsMsg'
Learn More

Starch indicator solution 1%, Acculute Standard Volumetric Solution, Final Volume 1L

Product Code. 11399329
Change view
Click to view available options
Quantity:
1 U
6 U
Unit Size:
1 unit
6 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399329 6 U 6 units
11389329 1 U 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399329 Supplier Thermo Scientific Alfa Aesar Supplier No. 088632.LS
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Iodometric Titrations | CAS: 9005-84-9

Starch indicator 1% solution is used as an indicator for redox titrations. It is also used in the detection of end point in iodometric titration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Starch indicator solution
CAS 9005-84-9, 7732-18-5
Physical Form Liquid,For iodometric titrations
Assay Percent Range 100%
MDL Number MFCD00082026
Solubility Information Miscible with water.
Quantity 6 U
Concentration or Composition (by Analyte or Components) Starch: 20%; Water: 80%
Vapor Pressure 17mm Hg at 20°C
What are the instructions for dilution of 1 unit of Starch indicator solution 1%, Acculute Standard Volumetric Solution, Final Volume 1L (Cat. No. 088632.LQ, 088632.LS)?

The following is the dilution protocol:
- Transfer entire contents of 1 unit bottle into a 1L volumetric flask.
- Rinse bottle residue, the threads of the bottle, and inner bottle cap each with DI water, and transfer to the 1L flask.
- Repeat rinse three times. Bring the 1L flask to volume with DI water and mix well.
The final solution in the 1L flask has a concentration of 1% Starch Indicator.

What is one unit of Starch indicator solution 1%, Acculute Standard Volumetric Solution, Final Volume 1L?

One unit of Starch indicator solution 1%, Acculute Standard Volumetric Solution, Final Volume 1 L makes one liter of 1% solution of starch indicator after dilution to one liter.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.