missing translation for 'onlineSavingsMsg'
Learn More

Starch, Extracted from Potato, MP Biomedicals™

Product Code. 11406372 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
2 kg
5 kg
Unit Size:
2kg
5kg
Product Code. Quantity unitSize
11406372 2 kg 2kg
11416372 5 kg 5kg
2 options
This item is not returnable. View return policy

Product Code. 11406372

Brand: MP Biomedicals 0210295402

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Starch is used as an additive for food processing, food starches are typically used as thickeners and stabilizers

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-25-8
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00082026,MFCD00132834
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
Synonym alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
PubChem CID 439341
ChEBI CHEBI:18167
IUPAC Name 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

Specifications

Physical Form Powder
Quantity 2 kg
Chemical Name or Material Starch
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.