missing translation for 'onlineSavingsMsg'
Learn More
Learn More
StAR Antibody (STAR/3976), Novus Biologicals™
Mouse Monoclonal Antibody
£178.00 - £351.00
Specifications
| Antigen | StAR |
|---|---|
| Clone | STAR/3976 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18657784
|
Novus Biologicals
NBP3-23661-20ug |
20 μg |
£178.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18619983
|
Novus Biologicals
NBP3-23661-100ug |
100 μg |
£351.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
StAR Monoclonal antibody specifically detects StAR in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| StAR | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| 10 mM PBS with 0.05% BSA | |
| 6770 | |
| IgG1 κ | |
| Protein A or G purified |
| STAR/3976 | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| cholesterol trafficker, mitochondrial steroid acute regulatory protein, StAR, StARD1, STARD1StAR-related lipid transfer (START) domain containing 1, START domain containing 1, START domain-containing protein 1, steroid acute regulatory protein, steroidogenic acute regulator, steroidogenic acute regulatory protein, steroidogenic acute regulatory protein, mitochondrial | |
| Recombinant fragment (around aa 39-108) of human StAR protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title