missing translation for 'onlineSavingsMsg'
Learn More
Learn More
StAR Antibody (STAR/3976) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24028
This item is not returnable.
View return policy
Description
StAR Monoclonal antibody specifically detects StAR in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| StAR | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry-Paraffin: 1-2 μg/mL | |
| cholesterol trafficker, mitochondrial steroid acute regulatory protein, StAR, StARD1, STARD1StAR-related lipid transfer (START) domain containing 1, START domain containing 1, START domain-containing protein 1, steroid acute regulatory protein, steroidogenic acute regulator, steroidogenic acute regulatory protein, steroidogenic acute regulatory protein, mitochondrial | |
| Recombinant fragment (around aa 39-108) of human StAR protein (exact sequence is proprietary) | |
| 100 μg | |
| Cancer | |
| 6770 | |
| Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| STAR/3976 | |
| Unconjugated | |
| 10 mM PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction