missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Staphylococcus Glu-C Recombinant Protein, PeproTech®
Click to view available options
Quantity:
1 mg
250 μg
50 μg
500 μg
Unit Size:
1mg
250µg
500µg
50µg
Description
Recombinant Staphylococcus Glu-C is a 28.8 kDa protease consisting of 266 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Proteases (also called Proteolytic Enzymes, Peptidases, or Proteinases) are enzymes that hydrolyze the amide bonds within proteins or peptides. Most proteases act in a specific manner, hydrolyzing bonds at, or adjacent to, specific residues, or a specific sequence of residues contained within the substrate protein or peptide. Proteases play an important role in most diseases and biological processes, including prenatal and postnatal development, reproduction, signal transduction, immune response, various autoimmune and degenerative diseases, and cancer. They are also an important research tool, as they are frequently used in the analysis and production of proteins. Glu-C cleaves at the Carboxyl side of E (can also cleave D under certain conditions).
Specifications
Specifications
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 66839245 |
| Molecular Weight (g/mol) | 28.8 kDa |
| Name | Staphylococcus Glu-C |
| Quantity | 1 mg |
| Source | E. coli |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Common Name | Glu-C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction