missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| For Use With (Application) | Immunoprecipitation |
| Formulation | Lyophilized from distilled water |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 42 kDa |
| Quantity | 5 mg |
| Storage Requirements | Store at -20°C. |
| Gene Alias | Ent A, EntA, Enterotoxin A, Enterotoxin type A, SEA, Staphylococcus enterotoxin type A |
| Reconstitution | Use sterile distilled water or physiological buffer of choice |
| Research Category | Virology Bacteria and Parasites |
| Purity or Quality Grade | >95%, by SDS-PAGE |
| Protein | Staphylococcus Enterotoxin |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?