missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Staphylococcus Enterotoxin B Monoclonal Antibody (2B33.5), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA546587
This item is not returnable.
View return policy
Description
Staphylococcus aureus Enterotoxin B consists of 239 amino acids with a molecular weight of ∽28 kDa.
Specifications
| Staphylococcus Enterotoxin B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| SEB | |
| 50 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| IgG1 |
| ELISA | |
| 2B33.5 | |
| Biotin | |
| Mouse | |
| Protein G | |
| RUO | |
| Bacteria | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction