missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Staphylococcus Enterotoxin A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109254
This item is not returnable.
View return policy
Description
Positive Control: Recombinant protein.
MA1-74347 recognises Staphylococcus aureus enterotoxin type A. Staphylococcus aureus produces several different types of enterotoxins (SE's). The SE's are short, soluble proteins that are highly stable and resistant to most proteolytic enzymes. They belong to a family of pyrogenic toxins and as such cause an increase in body temperature, immunosuppression and non-specific T-cell proliferation. These activities are referred to as superantigen activity and results from the interaction of SE's with T-cell antigen receptors and the major histocompatibility complex (MHC) on antigen presenting cells. In addition to their superantigen activity, SE's are also emetics (they cause vomiting). The SE's are important in food poisoning. Enterotoxin A is one of the SE's most commonly identified in clinical isolates. MA1-74347 may be used as a capture antibody in ELISA assays for S. aureus enterotoxin A in combination with MCA2848 as a detection agent.
Specifications
| Staphylococcus Enterotoxin A | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Recombinant protein within staphylococcus aureus Enterotoxin A aa 1-257. | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction