Learn More
Invitrogen™ Staphylococcus Enterotoxin A Monoclonal Antibody (D99G)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA17480
Description
The MA1-7480 antibody reacts with Staphylococcus Enterotoxin toxin A capture in bacterial and infected tissue samples. See related products for paired antibody. MA1-7480 has been successfully used in ELISA applications. The MA1-7480 antibody was raised against Staphylococcus Toxin A.
MA1-74347 recognises Staphylococcus aureus enterotoxin type A. Staphylococcus aureus produces several different types of enterotoxins (SE's). The SE's are short, soluble proteins that are highly stable and resistant to most proteolytic enzymes. They belong to a family of pyrogenic toxins and as such cause an increase in body temperature, immunosuppression and non-specific T-cell proliferation. These activities are referred to as superantigen activity and results from the interaction of SE's with T-cell antigen receptors and the major histocompatibility complex (MHC) on antigen presenting cells. In addition to their superantigen activity, SE's are also emetics (they cause vomiting). The SE's are important in food poisoning. Enterotoxin A is one of the SE's most commonly identified in clinical isolates. MA1-74347 may be used as a capture antibody in ELISA assays for S. aureus enterotoxin A in combination with MCA2848 as a detection agent.
Specifications
| Staphylococcus Enterotoxin A | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Staph. Toxin A. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
| ELISA | |
| D99G | |
| Unconjugated | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.