missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Staphylococcus Aureus Enterotoxin B Antibody (S643), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
£393.00
Specifications
| Antigen | Staphylococcus Aureus Enterotoxin B |
|---|---|
| Clone | S643 |
| Applications | Sandwich ELISA |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 532 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18569797
|
Novus Biologicals
NB100-73025AF532 |
0.2 mL |
£393.00
0.20mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Staphylococcus Aureus Enterotoxin B Monoclonal antibody specifically detects Staphylococcus Aureus Enterotoxin B in Bacteria samples. It is validated for ELISASpecifications
| Staphylococcus Aureus Enterotoxin B | |
| Sandwich ELISA | |
| Alexa Fluor 532 | |
| Mouse | |
| Bacteria | |
| S. aureus | |
| IgG1 | |
| Protein G purified |
| S643 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50mM Sodium Borate | |
| Hybridoma clone has been derived from hybridization of X63-Ag8-653 myeloma cells with spleen cells of Balb/c mice immunized with recombinant S. aureus enterotoxin B expressed in E. coli. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title