missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Staphylococcus aureus ALD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117600
This item is not returnable.
View return policy
Description
This antibody has been tested in direct-ELISA.
May play a role in cell wall synthesis as L-alanine is an important constituent of the peptidoglycan layer.
Specifications
| Staphylococcus aureus ALD1 | |
| Polyclonal | |
| Unconjugated | |
| Alanine dehydrogenase 1 | |
| Recombinant Staphylococcus aureus Alanine dehydrogenase 1 protein (1-372aa). | |
| 100 μg | |
| Primary | |
| -20°C or -80°C if preferred | |
| Liquid |
| Western Blot | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Rabbit | |
| Protein G | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction