missing translation for 'onlineSavingsMsg'
Learn More

Thermo Fisher Scientific™ Staphylococci aureus ATCC 9144 5pcs

Product Code. 12995923
Change view
Click to view available options
Quantity:
5 Loops
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12995923 5 Loops Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12995923 Supplier Thermo Fisher Scientific™ Supplier No. R4609321

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms

Recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.

The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY

Specifications

ATCC® ATCC - EULA required
Biosafety Level DG
Content And Storage 2°C to 8°C
Country of Origin US
Manufacturing Origin United States
Product Type Quality Control Organism
For Use With (Application) Ready-to-Use ATCC QC Organisms
No. per Pack 5
Packaging Type Foil Pk.
Quantity 5 Loops
Species Staphylococcus aureus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.