missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558427
This item is not returnable.
View return policy
Description
Immunogen sequence: ESGEEWRGFI LTVTELSVPQ NVSLLPGQVI KLHEVLEREK KRRIETEQST SVEKEKEPTE DYVDVLNPMP ACFYTVSRKE ATEMLQK Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 80%.
The protein encoded by this gene functions as a docking protein acting downstream of Tec tyrosine kinase in B cell antigen receptor signaling. The protein is directly phosphorylated by Tec in vitro where it participates in a postive feedback loop, increasing Tec activity.
Specifications
| STAP1 | |
| Polyclonal | |
| Unconjugated | |
| STAP1 | |
| AI586015; BCR downstream signaling 1; BCR downstream-signaling protein 1; BRDG1; docking protein BRDG1; signal transducing adaptor family member 1; signal-transducing adaptor protein 1; Stap1; STAP-1; Stem cell adaptor protein 1; stem cell adaptor protein STAP-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26228 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9ULZ2 | |
| STAP1 | |
| Recombinant protein corresponding to Human STAP1. Recombinant protein control fragment (Product #RP-96503). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction