missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STAP1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5149050
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
The protein encoded by this gene functions as a docking protein acting downstream of Tec tyrosine kinase in B cell antigen receptor signaling. The protein is directly phosphorylated by Tec in vitro where it participates in a postive feedback loop, increasing Tec activity.
Specifications
| STAP1 | |
| Polyclonal | |
| Unconjugated | |
| STAP1 | |
| AI586015; BCR downstream signaling 1; BCR downstream-signaling protein 1; BRDG1; docking protein BRDG1; signal transducing adaptor family member 1; signal-transducing adaptor protein 1; Stap1; STAP-1; Stem cell adaptor protein 1; stem cell adaptor protein STAP-1 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 26228 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9ULZ2 | |
| STAP1 | |
| E. coli-derived recombinant human BRDG1, Ala177-Ser280. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction