missing translation for 'onlineSavingsMsg'
Learn More

Stanozolol, Mikromol™

Product Code. 17964426
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17964426 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17964426 Supplier Mikromol Supplier No. MM1285.00
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Stanozolol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Stanozolol
CAS 10418-03-8
Analyte or Component Names Stanozolol
Shipping Condition Room Temperature
Molecular Formula C21 H32 N2 O
Certifications/Compliance ISO 17034
Synonym Stanozolol, 2'H-Androst-2-eno[3,2-c]pyrazol-17-ol, 17-methyl-, (5ÃŽ±,17ÃŽ2)-, 2'H-5ÃŽ±-Androst-2-eno[3,2-c]pyrazol-17ÃŽ2-ol, 17-methyl- (8CI), Cyclopenta[7,8]phenanthro[2,3-c]pyrazol-1-ol, 1,2,3,3a,3b,4,5,5a,6,7,10,10a,10b,11,12,12a-hexadecahydro-1,10a,12a-trimethyl- (6CI,7CI), (5ÃŽ±,17ÃŽ2)-17-Methyl-2'H-androst-2-eno[3,2-c]pyrazol-17-ol, 17-Methyl-5ÃŽ±-androstano[3,2-c]pyrazol-17ÃŽ2-ol, 17-Methyl-pyrazolo[4',3':2,3]-5ÃŽ±-androstan-17ÃŽ2-ol, 17ÃŽ±-Methyl-17ÃŽ2-hydroxy-5ÃŽ±-androstano(3,2-c)pyrazole, 17ÃŽ2-Hydroxy-17-methyl-5ÃŽ±-androstano[3,2-c]pyrazole, Anabol, Androstanazol, Androstanazole, Androstanazolestanazol, Estazol, NSC 233046, NSC 43193, Stanabolic, Stanazolol, Stromba, Strombaject, Tevabolin, Win 14833, Winstroid, Winstrol, Winstrol Depot, Winstrol V
InChI Formula InChI=1S/C21H32N2O/c1-19-11-13-12-22-23-18(13)10-14(19)4-5-15-16(19)6-8-20(2)17(15)7-9-21(20,3)24/h12,14-17,24H,4-11H2,1-3H3,(H,22,23)/t14-,15+,16-,17-,19-,20-,21-/m0/s1
SMILES C[C@]1(O)CC[C@H]2[C@@H]3CC[C@H]4Cc5[nH]ncc5C[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name (1S,2S,10S,13R,14S,17S,18S)-2,17,18-trimethyl-6,7-diazapentacyclo[11.7.0.0^{2,10}.0^{4,8}.0^{14,18}]icosa-4(8),5-dien-17-ol
Molecular Weight (g/mol) 328.49
Quantity 100 mg
Formula Weight 328.2515 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.