missing translation for 'onlineSavingsMsg'
Learn More

Stannous Chloride Dihydrate, ACS Reagent Grade, Ricca Chemical

Product Code. 31749821
Change view
Click to view available options
Quantity:
500 g
Packaging:
White Poly Wide Mouth Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
31749821 500 g White Poly Wide Mouth Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31749821 Supplier Ricca Chemical Company Supplier No. RDCS1010500B1
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 10025-69-1
Color Colorless
CAS Min % 98.0
CAS Max % 100.0
Physical Form Crystalline
Quantity 500 g
Packaging White Poly Wide Mouth Bottle
Molecular Weight (g/mol) 225.65 g/mol
Percent Purity 98 to 103%
Grade ACS
Chemical Name or Material Stannous Chloride Dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.