missing translation for 'onlineSavingsMsg'
Learn More

Stannous Chloride, 99%, (Crystalline), MP Biomedicals

Product Code. 11496602 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Unit Size:
100g
1kg
250g
Product Code. Quantity unitSize
11496602 100 g 100g
11406612 250 g 250g
11426612 1 kg 1kg
3 options
This item is not returnable. View return policy

Product Code. 11496602

Brand: MP Biomedicals 0215015780

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7772-99-8
Molecular Formula Cl2Sn
Molecular Weight (g/mol) 189.61
MDL Number MFCD00011241
InChI Key AXZWODMDQAVCJE-UHFFFAOYSA-L
PubChem CID 24479
ChEBI CHEBI:78067
IUPAC Name λ²-tin(2+) dichloride
SMILES [Cl-].[Cl-].[Sn++]

Specifications

Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99%
Formula Weight 189.6
Chemical Name or Material Stannous Chloride, Anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.