missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stanniocalcin 2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10314-1-AP-150UL
This item is not returnable.
View return policy
Description
Has an anti-hypocalcemic action on calcium and phosphate homeostasis.Specifications
| Stanniocalcin 2 | |
| Polyclonal | |
| Unconjugated | |
| STC2 | |
| stanniocalcin 2, Stanniocalcin related protein, STC 2, STC related protein, STC2, STCRP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8614 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O76061 | |
| STC2 | |
| Stanniocalcin 2 Fusion Protein Ag0359 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction