missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stanniocalcin 1/STC-1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97046AF750
This item is not returnable.
View return policy
Description
Stanniocalcin 1/STC-1 Polyclonal specifically detects Stanniocalcin 1/STC-1 in Human samples. It is validated for ELISA.
Specifications
| Stanniocalcin 1/STC-1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Alexa Fluor 750 | |
| stanniocalcin 1, stanniocalcin-1, STCSTC-1 | |
| Produced in rabbits immunized with purified, recombinant Stanniocalcin 1/STC-1 (NP_003146.1; Met1-Ala247). | |
| 0.1 mL | |
| Signal Transduction | |
| 6781 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction