missing translation for 'onlineSavingsMsg'
Learn More

Stanniocalcin 1/STC-1 Antibody (380715) [DyLight 680], Novus Biologicals™

Product Code. 30738043 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30738043 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30738043 Supplier Novus Biologicals Supplier No. FAB2958Y

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Stanniocalcin 1/STC-1 Monoclonal antibody specifically detects Stanniocalcin 1/STC-1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Stanniocalcin 1/STC-1
Applications Western Blot
Classification Monoclonal
Clone 380715
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias stanniocalcin 1, stanniocalcin-1, STCSTC-1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Stanniocalcin 1/STC-1, Thr18-Ala247, Accession # P52823
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6781
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.