Learn More
Scie Plas Standard Twin-Plate Maxi-Gel Electrophoresis Unit
Designed specifically for higher resolution PAGE techniques requiring self-cast gels.
Brand:  Scie Plas TV400Y
Additional Details : Weight : 1.30000kg
Product Code. 15805561
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
The unit of choice for maxi vertical gel electrophoresis. Glass plates are completely sealed against a soft silicone gasket ensuring no possibility of current leakage during extended runs.
Efficient cooling - glass plates are completely immersed in buffer, which acts as an effective heat sink, homogenising the gel temperature. The supercooled option further allows heat to be removed from the gel
  • Enhanced gaskets - ensure high resolution separations and no run time lag due to current leakage
  • Gel running module - allows unit to be safely picked up and transported around the laboratory
  • Hand grips on tank - allow for easy transportation around the lab. The ergonomic non slip grip ensures safe carrying even when loaded with glass plates, gels and buffer
  • Gel plate size 205mm x 200mm
Specifications
Specifications
Vertical Gel Unit
16.5W x 17.5 cm H
Running module glass plates spacers combs casting base