Learn More
Hach™ Addista Multi-Parameter Standard Solution
Product Code. 10663755
Change view
Click to view available options
For Use With (Application):
Orthophosphate, COD, Ammonium, Chloride, Nitrate, Phosphate (ortho), Sulphate, TOC
Sulphate, Ammonium, Chloride, COD, Nitrate, Phosphate (ortho), TOC
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) unitSize
10663755 Orthophosphate, COD, Ammonium, Chloride, Nitrate, Phosphate (ortho), Sulphate, TOC Each
10653755 Sulphate, Ammonium, Chloride, COD, Nitrate, Phosphate (ortho), TOC Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10663755 Supplier Hach™ Supplier No. LCA703
Make sure that analytical results are always correct. Hach™ Addista Multi-Parameter Standard Solution consists of one combined standard/spiking solution and two round robin test solutions. This system for water quality testing is used with Hach LCK cuvette tests.
With Hach Addista standard solutions, users can easily verify that their analytical approach to water quality testing (sampling, sample preparation, pipetting, photometric analysis) delivers correct results. Addista multiparameter solutions are designed for use with Hach LCK curvette tests.
Each kit comes with one standard solution and two solutions with an unknown concentration for participation in a free round-robin test. After successful participation in this quality assurance program, a certificate confirming the good quality of the analytical results is sent. If results are out of specifications, Hach experts will provide support in identifying and solving the issue.
  • Confidence in the results: Continuous documentation of Analytical Quality Assurance measures
  • Less complexity: Each solutions contains several parameters to reduce the number of single parameter standards in your lab
  • Fewer error sources: Ready-to use solution with Hach LCK cuvettes for less handling to eliminate error sources
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Orthophosphate, COD, Ammonium, Chloride, Nitrate, Phosphate (ortho), Sulphate, TOC
For Use With (Equipment) LCK curvette tests
Includes One standard solution and two solutions with an unknown concentration
Quantity 1 Kit
Type Standard Solutions