missing translation for 'onlineSavingsMsg'
Learn More

Standard Color Solution GY, Green/Yellow, EP Grade, Reagecon™
BIOPHARMA

Product Code. 11884232 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11884232 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11884232 Supplier Reagecon Supplier No. EP705

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used to calibrate, control, qualify and validate color measurement instruments. Standard Color Solution GY, Green/Yellow, EP Grade, Reagecon™ is a ready-to-use colorimetric solution produced in accordance with Eu. Ph. Chap 2 from a precise combination of Primary Color Solutions.

Ready-to-use colorimetric standard presented in a high-quality, tamper-evident bottle. EP Standard Color Solutions are prepared from the EP Primary Color Solutions (yellow, red and blue) and can be used to prepare EP Reference Color Solutions.
  • Manufactured according to European Pharmacopoeia Chapter 2 guidelines
  • Balance Certificate of Calibration issued in accordance with ISO 17025
  • Prepared gravimetrically on a weight/weight basis using OIML traceable weights
  • Concentration verified using a high-performance spectrophotometer calibrated using ISO Guide 34 standards
  • Color Standard GY = 9.6 Yellow + 0.2 Red + 0.2 Blue 

Specifications

Description European Pharmacopoeia Standard Color Solution GY
Packaging Tamper evident
Certificate of Analysis Yes
CAS 7705-08-0, 7646-79-9, 7758-98-7, 7647-01-0
Analyte or Component Names Iron chloride, Cobalt chloride, Copper sulfate, Hydrochloric acid
Physical Form Liquid
Color Green/Yellow
Element Iron Chloride, Cobalt Chloride, Copper Sulfate, Hydrochloric Acid
Quantity 125 mL
Grade Eu. Ph. Chapter 2
compliance-icons compliance-icons
Product Identifier
  • STANDARD SOLUTION GY (GREEN/YELLOW)
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H350i-May cause cancer by inhalation.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : iron trichloride, hydrochloric acid 37%,copper sulphate, cobalt dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.